EC HPLC column EC 150/2 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 150 mm, ID: 2 mm pack of 1 1 * 1 items
Catalog # MANA763436.20
Supplier: MACHEREY-NAGEL
EC HPLC column EC 150/2 NUCLEOSHELL Bluebird RP 18, 2.7 µm length: 150 mm, ID: 2 mm pack of 1 1 * 1 items
Catalog # MANA763436.20
Supplier: MACHEREY-NAGEL
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
HPLC columns, PFAS Delay, NUCLEODUR®
Supplier: MACHEREY-NAGEL
Organic reference standard, chlordecone, RS, Dr. Ehrenstorfer
Supplier: EHRENSTORFER