Order Entry
Italy
ContactUsLinkComponent
A KYN (Kynurenine) ELISA with a senstivity of 4.69pmol/ml and a range of 7.8-500pmol/ml. Uses the detection method Competitive ELISA , Coated with Antigen 1 * 1 KIT
Codice catalogo GENOIT8272
Supplier:  G-Biosciences
A KYN (Kynurenine) ELISA with a senstivity of 4.69pmol/ml and a range of 7.8-500pmol/ml. Uses the detection method Competitive ELISA , Coated with Antigen 1 * 1 KIT
Codice catalogo GENOIT8272
Supplier:  G-Biosciences
Frequently Bought Together