Order Entry
Italy
ContactUsLinkComponent
A Calprotectin (Calprotectin) ELISA with a senstivity of 4.688ng/ml and a range of 7.8-500ng/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8338
Supplier:  G-Biosciences
A Calprotectin (Calprotectin) ELISA with a senstivity of 4.688ng/ml and a range of 7.8-500ng/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8338
Supplier:  G-Biosciences
Frequently Bought Together