Order Entry
Italy
ContactUsLinkComponent
Immobilized Metal Ion Affinity Chromatography (IMAC), developed by Porath (1975), is based on the interaction of certain protein res idues (histidines, cysteines, and to some extent tryptophans) with cations of transition metals. 1 * 5 mL
Catalog # GENO786-1040
Supplier:  G-Biosciences
Immobilized Metal Ion Affinity Chromatography (IMAC), developed by Porath (1975), is based on the interaction of certain protein res idues (histidines, cysteines, and to some extent tryptophans) with cations of transition metals. 1 * 5 mL
Catalog # GENO786-1040
Supplier:  G-Biosciences
Frequently Bought Together