Order Entry
Italy
Orders LinkContactUsLinkComponent
Trypan blue solution 0,4%

Trypan blue solution 0,4%

Codice catalogo GENO786-1384
Nome del fornitore:   G-Biosciences
Trypan blue solution 0,4%
Codice catalogo GENO786-1384
Nome del fornitore:  G-Biosciences
Numero CAS:  72-57-1
Trypan blue solution 0,4%
Numero CAS:  72-57-1
Price
30,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

Trypan Blue 0.4% Solution is commonly used as a cell stain to measure cell viability using dye exclusion method. The dye exclusion method is based on the principle that cell impermeable dyes like Trypan blue will stain only dead cells where as viable cells will not be stained.

  • Solution is made in PBS, pH 7.4 ± 0.1 Sterile filtered Store at ambient temperature

Trypan Blue 0.4% Solution is routinely used for counting cells with hemocytometer during subculturing of cells. Trypan blue stain can be used to check cell viability accurately and quickly anytime.

Synonyms:  Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
  • CAS Number:72-57-1
  • Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
  • MDL Number:MFCD00003969
  • Molecular Weight:960.82 g/mol
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.