Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Trypan blue solution 0,4%

Catalog # GENO786-1383
Supplier:   G-Biosciences
Trypan blue solution 0,4%
Catalog # GENO786-1383
Supplier:  G-Biosciences
CAS Number:  72-57-1
Trypan blue solution 0,4%
CAS Number:  72-57-1
Price
16,10 €
Log in for availability
1 * 50 mL
Product Details & Documents

Trypan Blue 0.4% Solution is commonly used as a cell stain to measure cell viability using dye exclusion method. The dye exclusion method is based on the principle that cell impermeable dyes like Trypan blue will stain only dead cells where as viable cells will not be stained.

  • Solution is made in PBS, pH 7.4 ± 0.1 Sterile filtered Store at ambient temperature

Trypan Blue 0.4% Solution is routinely used for counting cells with hemocytometer during subculturing of cells. Trypan blue stain can be used to check cell viability accurately and quickly anytime.

Synonyms:  Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
  • CAS Number:72-57-1
  • Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
  • MDL Number:MFCD00003969
  • Molecular Weight:960.82 g/mol