Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
Italy
Inserisci ordini
Stato dell'ordine
Il tuo profilo
Carrello
ContactUsLinkComponent
NaCl Solution 2.50% AB250 (for ES-421[Cat.No4211], PAL-ES2, ES-3, SALT PROBE Calibration) 1 * 5 mL
Catalog # ATAGRE-120250
Supplier:
Atago
Expand image
NaCl Solution 2.50% AB250 (for ES-421[Cat.No4211], PAL-ES2, ES-3, SALT PROBE Calibration) 1 * 5 mL
Catalog # ATAGRE-120250
Supplier:
Atago
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW