Order Entry
Italy
ContactUsLinkComponent
Complete 6mm Fiber Optic Dip Probe System. For GENESYS 150 & 180. Includes: Fiber optic coupler insert for sample compartment, 6m m Stainless steel transflectance probe with interchangeable tip, 10mm tip for probe (pre-attached to probe body), Printe 1 * 1 items
Catalog # 634-1104
Complete 6mm Fiber Optic Dip Probe System. For GENESYS 150 & 180. Includes: Fiber optic coupler insert for sample compartment, 6m m Stainless steel transflectance probe with interchangeable tip, 10mm tip for probe (pre-attached to probe body), Printe 1 * 1 items
Catalog # 634-1104
Frequently Bought Together