Order Entry
Italy
ContactUsLinkComponent
Cd274 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of mouse Cd274 in cell cult ure supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5130
Supplier:  Abnova
Cd274 (Mouse) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of mouse Cd274 in cell cult ure supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5130
Supplier:  Abnova
Frequently Bought Together