Order Entry
Italy
ContactUsLinkComponent
CD274 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human CD274 in cell cult ure supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5129
Supplier:  Abnova
CD274 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human CD274 in cell cult ure supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5129
Supplier:  Abnova
Frequently Bought Together