Securely support a laptop computer within easy view of operator. Mounts to front leg of SoLo Hydraulic Base Stand. Raises, lowers and pivots. Includes powder-coated steel arm, support surface and mounting hardware. Installation is required.EA 1 * 1 items
Catalog # LBCP3869200
Supplier: LABCONCO
Frequently Bought Together