TBN STANDARD 10 MG/G KOH 1 * 50 g
Catalog # REGERETBN10
Supplier: REAGECON
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
IBUPROFEN FOR PEAK IDENTIFICATION 1 * 0,09 mg
Supplier: EDQM
Accessories for ion selective meters, LAQUAtwin
Supplier: HORIBA
Pentabromophenol
Supplier: EHRENSTORFER
Buffer solution pH 7, Phosphate buffer
Supplier: Thermo Fisher Scientific
Tri(2-ethylhexyl) trimellitat 1 * 100 mg
Supplier: EHRENSTORFER
alpha-HCH D6 certified 1 * 10 mg
Supplier: EHRENSTORFER
200g 1 * 200 g
Supplier: LGC Standards PROMOCHEM
Mancozeb certified 1 * 100 mg
Supplier: EHRENSTORFER
Dimethyl fumarate
Supplier: EHRENSTORFER