1,6-Naphthyridine
Bottle, 1 g
Catalog # APOSOR42177-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Pyrido[4,3-b]pyridine, 1,6-Diazanaphthalene
- CAS Number:253-72-5
- Formula:C₈H₆N₂
- MDL Number:MFCD00059750
- Molecular Weight:130.15 g/mol
- Melting Pt:35 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



