SeramunBlock
SeramunBlock, 100 mL
Catalog # APOSBIL3122-100ML
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Combined Stabiliser/ blocking solution for solid phase immobilised proteins effecting protein stabilisation and blocking of free surfaces.
Specifications
Specifications
- Pk:100 mL
- Description:SeramunBlock