Caps from 2202-9125 - 13mm PP cap. Process: Particulate Cleaning & Autoclave. Packaging : Single autoclave bag each cap. Certification: C of A Endotoxin (Report Actual Results), C of A USP Particles, C of A USP Sterility, Certificate of Processing, & 1 * 72 items
Catalog # NALGCEE6MM13PA-E1
Supplier: Thermo Fisher Scientific
Frequently Bought Together