Order Entry
Italy
ContactUsLinkComponent
Contains a proprietary mixture of proteins and buffer reagents designed to reduce heterophilic interactions in ELISA assays utilizing mouse immunoreagents. 1 * 1.250 µG
Catalog # BSENBL-002-1250
Supplier:  Biosensis
Contains a proprietary mixture of proteins and buffer reagents designed to reduce heterophilic interactions in ELISA assays utilizing mouse immunoreagents. 1 * 1.250 µG
Catalog # BSENBL-002-1250
Supplier:  Biosensis
Frequently Bought Together