Order Entry
Italy
ContactUsLinkComponent
Tenacity No. 125 Paste, a high temperature flux paste, working range 800-1100C (1472-2012F) 1 * 700 g
Catalog # 47289.39
Tenacity No. 125 Paste, a high temperature flux paste, working range 800-1100C (1472-2012F) 1 * 700 g
Catalog # 47289.39
Frequently Bought Together