Order Entry
Italy
ContactUsLinkComponent
pHG311-9 - pH Electrode, Ag/AgCl, S7 screw cap, for alkaline samples pH 0…14, 0…80°C, 12 mm x 103 mm 1 * 1 items
Catalog # HACHE11M004
Supplier:  Hach
pHG311-9 - pH Electrode, Ag/AgCl, S7 screw cap, for alkaline samples pH 0…14, 0…80°C, 12 mm x 103 mm 1 * 1 items
Catalog # HACHE11M004
Supplier:  Hach
Frequently Bought Together