Advanced MB, InduProof®
TKG-104-MB-IP68-VESA-GREY-USB, DE, with mousebutton, 340×165×16,3 mm
Catalog # GETTKG17203
Supplier: GETT GERATETECHNIK
Jump to:
Product Details & Documents
Rugged, water- and dust-proof silicone keyboard with integrated mouse button.
- Rugged, durable keyboard with 104 keys and integrated mouse button
- Comfortable & ergonomic operation also with protective gloves made of silicone or latex
- IP 68 protection, can be submerged into liquids
- Easy cleaning and disinfection - even under tap water or by immersion
- Increased chemical and mechanical resistance due to special surface treatment
- VESA-M5-boreholes (distance 75×75 mm)
- 4 years warranty on technical function
- Made in Germany
The InduProof® Advanced has a full set of keys with a numeric keyboard, an integrated mouse button (mini-joystick) and is a convincing product thanks to its pleasant sense of touch and the very high degree of protection that it provides. As a result of its robust design, it is highly suitable for use in challenging, hygiene-sensitive areas like industrial food and drinks production. It is possible to completely clean and disinfect the unit. It has a long serviceable life as a result of its high-quality workmanship – after all it is 'Made in Germany'.
: CE, REACH, RoHs, EMC, IP 68.
: The keyboards are packed in brown cardboard boxes.
Specifications
- Description:TKG-104-MB-IP68-VESA-GREY-USB, DE, with mousebutton
- W×D×H:340×165×16,3 mm



