SPE columns, CHROMABOND® Easy
Easy, PP, 50 Å, 80 µm, 6 ml, 200 mg
Catalog # MANA730755.250
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
CHROMABOND® Easy is a polar, bifunctionally modified polystyrene-divinylbenzene copolymer for SPE.
- Applicable for a wide range of analytes without preconditioning
- More hydrophilic than conventional polystyrene-divinylbenzene polymers
- Easily wettable with water
Applications: Polar pesticides (acidic, neutral, basic) or phenols from water; drug analysis from urine, blood, serum or from tablets and creams.
Specifications
- Particle Size:80 µm
- Pore size:50 Å
- Capacity:6 ml
- Seq:08
- Bed weight:200 mg
- Column hardware:PP
- Phase:Easy
- Cat. No.:MANA730755.250
- pH range:1 - 14
- Surface area:650 - 700 m²/g



