Replacement flow measuring cell inclusive measuring cell glass 4 mm and 12 mm cpl. for inserting into the flow measuring cell of the wet dispersion unit article no. 22.8500.00 for Laser Particle Sizers ANALYSETTE 22 MicroTec plus and NanoTec plus 1 * 1 items
Catalog # FRIT22.8560.04
Supplier: FRITSCH
Frequently Bought Together