Order Entry
Italy
ContactUsLinkComponent
With the screw-on grinding bowl lid made of zirconium oxide, you can seal the zirconium oxide grinding bowl with 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between fi lling in the glove box and the mil 1 * 1 items
Catalog # FRIT50.6495.00
Supplier:  FRITSCH
With the screw-on grinding bowl lid made of zirconium oxide, you can seal the zirconium oxide grinding bowl with 500 ml, 250 ml and 150 ml volume of the PULVERISETTE 5 premium line gas-tight for transport between fi lling in the glove box and the mil 1 * 1 items
Catalog # FRIT50.6495.00
Supplier:  FRITSCH
Frequently Bought Together