Order Entry
Italy
ContactUsLinkComponent
The rotor made of hardened stainless steel with straight cutting edges which run parallel to the fixed knives is suitable for easil y cut material, such as hay, straw, fibrous or biological materials. The rotor knives may be turned, since they have 1 * 1 items
Catalog # FRIT45.5185.00
Supplier:  FRITSCH
The rotor made of hardened stainless steel with straight cutting edges which run parallel to the fixed knives is suitable for easil y cut material, such as hay, straw, fibrous or biological materials. The rotor knives may be turned, since they have 1 * 1 items
Catalog # FRIT45.5185.00
Supplier:  FRITSCH
Frequently Bought Together