Order Entry
Italy
ContactUsLinkComponent
test sieve 400/65 mm mesh = 70 = 212 µm - ASTM - E-11-1995 - mesh 1 * 1 items
Catalog # FRIT34.4610.02
Supplier:  FRITSCH
test sieve 400/65 mm mesh = 70 = 212 µm - ASTM - E-11-1995 - mesh 1 * 1 items
Catalog # FRIT34.4610.02
Supplier:  FRITSCH
Frequently Bought Together