Order Entry
Italy
Orders LinkContactUsLinkComponent
Cleaning cloths, MicroClean Plus

Cleaning cloths, MicroClean Plus

Green, 400×450 mm, 5 cloths/pack
Catalog # VILE152533
Supplier:   FHP VILEDA SCS
Cleaning cloths, MicroClean Plus
Catalog # VILE152533
Supplier:  FHP VILEDA SCS
Cleaning cloths, MicroClean Plus
Cleaning cloths, MicroClean Plus

Green, 400×450 mm, 5 cloths/pack

Price
621,00 €
Log in for availability
1 * 100 pezzi
Product Details & Documents

MicroClean Plus is a high performance cleaning cloth made from a uniquely fine microfibre (70% polyester and 
30% polyamide). Great streak- and lint-free results are achieved with just one wipe. Ideal for quick dirt removal even without chemicals.

  • Extremly durable
  • Micron Fibre Technology for deeper and more effective cleaning
  • MicroClean dries quickly: Bacteria growth is prevented
  • Washable and reusable: Reduces waste and protects the environment
  • Colour coding system allows a cleaning process according to hygiene standards

MicroClean Plus is suitable for demanding cleaning tasks in general building cleaning and Hotels/Restaurants/Catering on every kind of surface - with or without cleaning agents.

Specifications
  • Packed:
    5 cloths/pack
  • Colour:
    Green
  • L×W:
    400×450 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.