Order Entry
Italy
ContactUsLinkComponent
KIT ANTIROTATION PIPE PRECELLYS Sparepart for Precellys 24 and Precellys Dual 24 Recommanded frequency replacement : 1 year This spa re parts can be replaced by the user as described in the user manual 1 * 1 items
Catalog # 432-0243
Supplier:  Bertin Instruments
KIT ANTIROTATION PIPE PRECELLYS Sparepart for Precellys 24 and Precellys Dual 24 Recommanded frequency replacement : 1 year This spa re parts can be replaced by the user as described in the user manual 1 * 1 items
Catalog # 432-0243
Supplier:  Bertin Instruments
Frequently Bought Together