Order Entry
Italy
ContactUsLinkComponent
Optical drive for ImageXpress Micro 4 including >10,000 hour solid state light source, large sensor scientific CMOS detector and opt ical train to accommodate large field of view imaging. 1 * 1 items
Catalog # MLDV5046687
Supplier:  Molecular Devices
Optical drive for ImageXpress Micro 4 including >10,000 hour solid state light source, large sensor scientific CMOS detector and opt ical train to accommodate large field of view imaging. 1 * 1 items
Catalog # MLDV5046687
Supplier:  Molecular Devices
Frequently Bought Together