Order Entry
Italy
Orders LinkContactUsLinkComponent
High content cell imaging system, ImageXpress Micro 4
High content cell imaging system, ImageXpress Micro 4
Filter set, Cy3
Catalog # MLDV5039858
Supplier:   Molecular Devices
High content cell imaging system, ImageXpress Micro 4
Catalog # MLDV5039858
Supplier:  Molecular Devices
High content cell imaging system, ImageXpress Micro 4
High content cell imaging system, ImageXpress Micro 4
Filter set, Cy3
Price
Inquire For Price
Log in for availability
1 * 1 pezzi
Product Details & Documents
The ImageXpress Micro 4 high content imaging system represents the culmination of four generations of imaging expertise. The agile design allows you to boost your research with this faster than ever system, while providing the option to upgrade to confocal in the future to align with your research needs.
  • Tackle a wide range of assays with oil and air objectives ranging from 1× to 100×
  • Automated five-position filter cube changer to facilitate multiplex assays with up to five filter sets and four objectives for a single experiment
  • Optically-encoded linear voice coil driven stages with encoders that measure position with better than 25 nm resolution, to achieve high accuracy and precision in X, Y and Z axes
  • Expand your research capabilities with environmental control: Temperature, humidity and CO₂ control for multi-day, live cell experiments), transmitted light true phase contrast, confocal imaging (field upgradable), and fluidics (onboard disposable pipetting)
Capture images of whole organisms and cellular or intracellular events with the ImageXpress Micro 4 system, configured to suit your specific biological needs. We offer unlimited configurability with user exchangeable filter cubes, a wide range of objective lenses and environmental control, transmitted light, confocal imaging and fluidic options. Speed up your research by acquiring more than 245000 wells per day at 4× magnification with the system’s unique multi-well crop feature, and resolve the fine detail you need with increasing magnifications.
Combined with MetaXpress high content image acquisition and analysis software, the ImageXpress Micro 4 system is an end-to-end solution to interpret images, understand data and explore ideas.
Specifications
  • Description:
    Filter set, Cy3
  • Seq:
    106
Complementary Products
Product Information
  • Catalog #
    Description
    Pk
    Serum format
    Sterility
    Packed
    Recommended working volume
    Culture area
    Sterile
    Colour
    Working volume
    Availability
    Price
  • 30026.
    Calcein-AM Cell Viability Assay kit, 1000 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 417,00 €
  • 30029.
    NucView™ 488 Caspase-3 Assay Kit for live cells, 100 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 631,00 €
  • S1810-500
    Foetal bovine serum, South American origin
    Frozen
    15
    Log in for availability
    1 * 500 mL - 322,00 €
  • 100.103.010
    GrowDex®, 10 ml syringe
    1
    Log in for availability
    1 * 1 pezzi - 290,00 €
  • 30002.
    Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 440,00 €
  • 30009-2
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    1 KIT
    Log in for availability
    1 * 1 KIT - 294,00 €
  • 734-2327
    1/bag
    0,075 - 0,20 ml
    0,33 cm²
    +
    17
    Clear
    Log in for availability
    1 * 100 pezzi - 213,00 €
  • 735-0523
    Microlite™ 1+ strip, 1×12 per strip
    Log in for availability
    1 * 320 pezzi - 228,00 €
  • CORN3904BC
    20/bag
    96-well microplate, clear flat bottom, tissue culture treated, with lid
    75 - 200 µl
    21
    360 µl
    Black
    Log in for availability
    1 * 100 pezzi - 1 160,00 €
  • 734-2097
    200 µl
    96-MicroWell™ plate, flat bottomed wells, with lid
    Transparent
    Log in for availability
    1 * 50 pezzi - 221,00 €