Order Entry
Italy
ContactUsLinkComponent
Set Precision-therm., similar ISO 655, long, -35+600°C, enclosed scale, 6 therm., in stuffed case, suitable for government verificat ion respectively suitable for government calibration 1 * 1 items
Codice catalogo AMARN65122-SET
Supplier:  Amarell
Set Precision-therm., similar ISO 655, long, -35+600°C, enclosed scale, 6 therm., in stuffed case, suitable for government verificat ion respectively suitable for government calibration 1 * 1 items
Codice catalogo AMARN65122-SET
Supplier:  Amarell
Frequently Bought Together