Order Entry
Italy
ContactUsLinkComponent
Therm. similar Landsberger, +67+85:0,05°C, lower part 180x6-7mm, enclosed scale, capillary tube specially coated inside, with non-w etting blue special liquid, 160mm immersion, 480mm long, Richter-top finish with button, null
Codice catalogo AMARN63958-CB/CC
Supplier:  Amarell
Therm. similar Landsberger, +67+85:0,05°C, lower part 180x6-7mm, enclosed scale, capillary tube specially coated inside, with non-w etting blue special liquid, 160mm immersion, 480mm long, Richter-top finish with button, null
Codice catalogo AMARN63958-CB/CC
Supplier:  Amarell
Frequently Bought Together