Order Entry
Italy
ContactUsLinkComponent
Set Anschütztherm., -10+360°C:0,1°C, solid stem, white backed, 7 therm., in stuffed case, suitable for government calibration 1 * 1 items
Catalog # AMARN60086-SET
Supplier:  Amarell
Set Anschütztherm., -10+360°C:0,1°C, solid stem, white backed, 7 therm., in stuffed case, suitable for government calibration 1 * 1 items
Catalog # AMARN60086-SET
Supplier:  Amarell
Frequently Bought Together