Order Entry
Italy
ContactUsLinkComponent
Set Therm., similar Dr. Otte, similar DIN, 0+580°C, enclosed scale, 5 therm., in stuffed case, suitable for government verification respectively suitable for government calibration 1 * 1 items
Catalog # AMARN59820-SET
Supplier:  Amarell
Set Therm., similar Dr. Otte, similar DIN, 0+580°C, enclosed scale, 5 therm., in stuffed case, suitable for government verification respectively suitable for government calibration 1 * 1 items
Catalog # AMARN59820-SET
Supplier:  Amarell
Frequently Bought Together