Order Entry
Italy
ContactUsLinkComponent
Softening point therm., similar DIN 12785, +30+200:0,5°C, similar ASTM 16 C/IP 61 C, solid stem, white backed, red special liquid, t otal immersion, 395x7mm, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARM255102-FL
Supplier:  Amarell
Softening point therm., similar DIN 12785, +30+200:0,5°C, similar ASTM 16 C/IP 61 C, solid stem, white backed, red special liquid, t otal immersion, 395x7mm, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARM255102-FL
Supplier:  Amarell
Frequently Bought Together