Order Entry
Italy
ContactUsLinkComponent
Thermostat control therm., -5+100:0,2°C, milk glass scale, capillary tube specially coated inside, with non-wetting blue special li quid, upper part 350mm, diameter throughout 9mm, immersion 140mm, Richter-top finish without button, null
Catalog # AMARM246214-CB
Supplier:  Amarell
Thermostat control therm., -5+100:0,2°C, milk glass scale, capillary tube specially coated inside, with non-wetting blue special li quid, upper part 350mm, diameter throughout 9mm, immersion 140mm, Richter-top finish without button, null
Catalog # AMARM246214-CB
Supplier:  Amarell
Frequently Bought Together