Angle stem therm., similar DIN 16175, -30+50:1°C, cap. prismatic, yellow backed, milk glass scale, red special liquid, upper part 22 8x20,5mm, lower part 125x8mm, lower part bent behind at 90° angle 1 * 1 items
Catalog # AMART96196-FL
Supplier: Amarell
Frequently Bought Together