Order Entry
Italy
ContactUsLinkComponent
Angle stem therm., similar DIN 16175, 0+100:1°C, cap. prismatic, yellow backed, milk glass scale, red special liquid, upper part 228 x20,5mm, lower part 185x8mm, lower part bent behind at 90° angle 1 * 1 items
Catalog # AMART96254-FL
Supplier:  Amarell
Angle stem therm., similar DIN 16175, 0+100:1°C, cap. prismatic, yellow backed, milk glass scale, red special liquid, upper part 228 x20,5mm, lower part 185x8mm, lower part bent behind at 90° angle 1 * 1 items
Catalog # AMART96254-FL
Supplier:  Amarell
Frequently Bought Together