AMA Freezer therm., complete in plastic casing, -35+20:0,5C, c. 280x28mm, enclosed scale, cap. yellow backed, red alcohol filled, rounded top finish, government verified with Verification Stamp null
Catalog # AMART108406-FL/VS
Nome del fornitore: Amarell
Frequently Bought Together