Order Entry
Italy
ContactUsLinkComponent
Precision therm., similar ISO 656, short, +58+82:0,2°C, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max. 250x9mm, richter-top finish with button, with ice point scale, null
Catalog # AMARN65750-CB
Supplier:  Amarell
Precision therm., similar ISO 656, short, +58+82:0,2°C, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max. 250x9mm, richter-top finish with button, with ice point scale, null
Catalog # AMARN65750-CB
Supplier:  Amarell
Frequently Bought Together