Order Entry
Italy
ContactUsLinkComponent
Thermometer similar to S.T.P.T.C. T 5d, solid stem, white backed, 15+45:0,1°C, length 430mm, imm. 100mm, capillary tube specially c oated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400030-CB
Supplier:  Amarell
Thermometer similar to S.T.P.T.C. T 5d, solid stem, white backed, 15+45:0,1°C, length 430mm, imm. 100mm, capillary tube specially c oated inside, with non-wetting blue special liquid, durable pigment, null
Catalog # AMARA400030-CB
Supplier:  Amarell
Frequently Bought Together