Order Entry
Italy
ContactUsLinkComponent
Set Hydrometers for plants, DIN 12792 (old), 0,610-2,000:0,002g/cm , ref. temp. 20°C, without therm., 9 instruments, in stuffed case , suitable for government verification 1 * 1 items
  AMARH820270
 :  Amarell
Set Hydrometers for plants, DIN 12792 (old), 0,610-2,000:0,002g/cm , ref. temp. 20°C, without therm., 9 instruments, in stuffed case , suitable for government verification 1 * 1 items
  AMARH820270
 :  Amarell
Frequently Bought Together