Order Entry
Italy
ContactUsLinkComponent
Set Hydrometers, DIN 12791/BS 718, L50, 0,60-2,00, ref. temp. 20°C, without therm., 28 hydrometers, in stuffed case, suitable for go vernment verification 1 * 1 items
Codice catalogo AMARH806260
Supplier:  Amarell
Set Hydrometers, DIN 12791/BS 718, L50, 0,60-2,00, ref. temp. 20°C, without therm., 28 hydrometers, in stuffed case, suitable for go vernment verification 1 * 1 items
Codice catalogo AMARH806260
Supplier:  Amarell
Frequently Bought Together