Order Entry
Italy
ContactUsLinkComponent
Thermostat control therm., +149+201:0,1°C, milk glass scale, cap. prismatic, colourless, gallium filled, upper part 350mm, diameter throughout 9mm, immersion 140mm, Richter-top finish without button, suitable for government calibration 1 * 1 items
Catalog # AMARM246210-GA
Supplier:  Amarell
Thermostat control therm., +149+201:0,1°C, milk glass scale, cap. prismatic, colourless, gallium filled, upper part 350mm, diameter throughout 9mm, immersion 140mm, Richter-top finish without button, suitable for government calibration 1 * 1 items
Catalog # AMARM246210-GA
Supplier:  Amarell
Frequently Bought Together