Order Entry
Italy
ContactUsLinkComponent
Viscometer therm. designed for the ball-draw viscometer, similar DIN 12785, +199+251:0,1°C, enclosed scale, cap. prismatic, colourle ss, gallium filled, immersion 140mm, Richter-top finish without button, 460x9,5mm, suitable for government calibratio 1 * 1 items
Catalog # AMARM241046-GA
Supplier:  Amarell
Viscometer therm. designed for the ball-draw viscometer, similar DIN 12785, +199+251:0,1°C, enclosed scale, cap. prismatic, colourle ss, gallium filled, immersion 140mm, Richter-top finish without button, 460x9,5mm, suitable for government calibratio 1 * 1 items
Catalog # AMARM241046-GA
Supplier:  Amarell
Frequently Bought Together