Order Entry
Italy
ContactUsLinkComponent
Set Density hydrometer, 0,600-2,000:0,001g/cm , ref. temp. 20°C, with therm., 0+40:1°C, 350mm long, special liquid, 14 hydrometers, in stuffed case 1 * 1 items
Catalog # AMARH801240-SET
Supplier:  Amarell
Set Density hydrometer, 0,600-2,000:0,001g/cm , ref. temp. 20°C, with therm., 0+40:1°C, 350mm long, special liquid, 14 hydrometers, in stuffed case 1 * 1 items
Catalog # AMARH801240-SET
Supplier:  Amarell
Frequently Bought Together