Soil therm., similar DIN 58655, -20+40:0,2°C, built-in length 110mm, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 345x18mm, obtuse angle of 150° bent behind, with swelling, null
Catalog # AMARK188006-CB
Supplier: Amarell
Frequently Bought Together