Soil therm., similar DIN 58655, -25+45:0,2°C, built-in length 60mm, milk glass scale, capillary tube specially coated inside, with non-wetting blue special liquid, upper part 345x18mm, obtuse angle of 150° bent behind, with swelling, null
Catalog # AMARK188004-CB
Supplier: Amarell
Frequently Bought Together