Order Entry
Italy
ContactUsLinkComponent
General purpose therm., solid stem, -10+360:2C, 300x6-7mm, Eco-friendly, green filled, biodegradable, white backed, immersion 76mm, with ring at top, in transparent square plastic case 1 * 1 items
Catalog # AMARG11408-EF
Supplier:  Amarell
General purpose therm., solid stem, -10+360:2C, 300x6-7mm, Eco-friendly, green filled, biodegradable, white backed, immersion 76mm, with ring at top, in transparent square plastic case 1 * 1 items
Catalog # AMARG11408-EF
Supplier:  Amarell
Frequently Bought Together