Order Entry
Italy
ContactUsLinkComponent
Precision therm., low temp., similar DIN, -38+50:0,2C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 400x6-7mm, with ring at top, null
Catalog # AMARL33102-CB
Supplier:  Amarell
Precision therm., low temp., similar DIN, -38+50:0,2C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 400x6-7mm, with ring at top, null
Catalog # AMARL33102-CB
Supplier:  Amarell
Frequently Bought Together