Order Entry
Italy
ContactUsLinkComponent
Precision therm., high temp., similiar DIN, +20+610:1°C, enclosed scale, cap. lens front, colourless, total immersion, gallium fille d, 450mm long, diameter 9-10mm, durable pigment, richter-top finish, with ice point scale, suitable for government ve 1 * 1 items
Catalog # AMARL29726-GA
Supplier:  Amarell
Precision therm., high temp., similiar DIN, +20+610:1°C, enclosed scale, cap. lens front, colourless, total immersion, gallium fille d, 450mm long, diameter 9-10mm, durable pigment, richter-top finish, with ice point scale, suitable for government ve 1 * 1 items
Catalog # AMARL29726-GA
Supplier:  Amarell
Frequently Bought Together