Thermometer, solid stem, similar to ASTM 46 F, white backed, 119,5+124,5:0,1F, capillary tube specially coated inside, with non-we tting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA300660-CB
Supplier: Amarell
Frequently Bought Together